missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA599293
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ELOVL1.
Elongation of very long chain fatty acid-like (ELOVL) proteins 1-6 are members of the ELO family of proteins, which play an important role in tissue-specific biosynthesis of very long chain fatty acids and sphingolipids. The ELOVL proteins act as catalysts in fatty acid elongation reduction and localize to the endoplasmic reticulum (ER). Elongation of very long chain fatty acids protein 1 (ELOVL1), also referred to as Ssc1, is the human homolog of the yeast ELO3 protein. It is expressed in a variety of tissues and at especially high levels in stomach, skin, intestine, kidney and lung. ELOVL1 participates in the elongation of very long chain saturated and monounsaturated fatty acids of up to 26 carbons and may be required for the development of a barrier in epithelial cells and skin. ELOVL1 is also important for the formation of Myelin in the central nervous system. Impaired ELOVL1 activity may be associated with disorders of sphingolipid metabolism.
Specifications
ELOVL1 | |
Polyclonal | |
Unconjugated | |
ELOVL1 | |
3-keto acyl-CoA synthase ELOVL1; AA407424; BB151133; CGI88; CGI-88; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1; elongation of very long chain fatty acids protein 1; ELOVL FA elongase 1; ELOVL fatty acid elongase 1; ELOVL1; sequence similarity to Cig30 1; Ssc1; Very long chain 3-ketoacyl-CoA synthase 1; very long chain 3-oxoacyl-CoA synthase 1; very-long-chain 3-oxoacyl-CoA synthase 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54325, 64834, 679532 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BW60, Q9JLJ5 | |
ELOVL1 | |
A synthesized peptide derived from human ELOVL1(Accession Q9BW60), corresponding to amino acid residues V108-I158. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELOVL1 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction