Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand: Merck MAHAS4510
This item is not returnable.
View return policy
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Standard | |
Mixed Cellulose Esters | |
0.45 μm | |
Clear | |
Polystyrene | |
Sterile | |
ELISPOT Assays |
Yes | |
8 x 12 | |
MultiScreen | |
MultiScreen-HA | |
96 | |
0.28 cm2 | |
10/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Product Title
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays > Multiscreen-HA; ELISPOT; Styrene; Mixed cellulose esters; 0.45μm; Clear; 10/Pk.
Spot an opportunity for improvement?Share a Content Correction