Learn More
Fisherbrand™ EPA Screw Neck Vial, Glass
Broad range of EPA Vials in clear and amber glass.
44.35 EUR - 90.05 EUR
Specifications
Description | Screw Neck |
---|---|
Diameter (Metric) Outer | 27.5 mm |
Material | Glass |
For Use With (Application) | TOC (EPA) |
Thread Style | 24-400 |
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
10465982
|
Fisherbrand™
24 09 0402 |
40 mL | Clear |
63.45 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
10000782
|
Fisherbrand™
24 09 0589 |
20 mL | Clear |
44.35 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
10758874
|
Fisherbrand™
24 09 0839 |
30 mL | Clear |
65.60 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
10458082
|
Fisherbrand™
24 09 0927 |
20 mL | Amber |
90.05 EUR
Pack of 100 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||||
11510585
|
Fisherbrand™
24 09 0923 |
30 mL | Amber |
84.05 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
11530585
|
Fisherbrand™
24 09 0928 |
40 mL | Amber |
51.80 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
11540585
|
Fisherbrand™
24 09 1089 |
60 mL | Clear |
79.20 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
11550585
|
Fisherbrand™
24 09 1090 |
60 mL | Amber |
82.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- EPA Vials can be delivered with a certificate of cleanliness
- EPA Vials can be obtained with any type of Screw Seal ND24 already screwed on
- Clear and amber glass
- Available in 20mL, 30mL, 40mL and 60mL
Specifications
Screw Neck | |
Glass | |
24-400 |
27.5 mm | |
TOC (EPA) |