missing translation for 'onlineSavingsMsg'
Learn More

Epithelial Membrane Antigen/EMA Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-348-P1ABX

 View more versions of this product

Product Code. 12631107

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown to be useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.

Host Species: Mouse

Clone: GP1.4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: DTRP

Immunogen: Human milk fat globule membranes

Molecular Weight: 265-400 kDa

Positive Control: Breast or Colon CA

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Epithelial Membrane Antigen/EMA Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Oncogenesis
Human
IgG1
Immunohistochemistry (Paraffin)
GP1.4
Unconjugated
Human milk fat globule membranes
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.