Learn More
Esonarimod, MedChemExpress
MedChemExpress Esonarimod (KE-298) is an antirheumatic agent.

POA - 515.70 EUR
Specifications
Chemical Name or Material | Esonarimod |
---|---|
Physical Form | Solid |
CAS | 101973-77-7 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16487768
![]() |
Medchem Express
HY-19440 |
10 mg |
97.20 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16487616
![]() |
Medchem Express
HY-19440 |
100 mg |
515.70 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16443737
![]() |
Medchem Express
HY-19440 |
50 mg |
321.30 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430238
![]() |
Medchem Express
HY-19440 |
5 mg |
70.20 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401819
![]() |
Medchem Express
HY-19440 |
1 mg |
POA
1mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Esonarimod (KE-298) is an antirheumatic agent.Specifications
Esonarimod | |
101973-77-7 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 100 mg/mL (356.71 mM; Need ultrasonic) | |
280.34 | |
99.94% |
Solid | |
White | |
C14H16O4S | |
KE-298 | |
CC1=CC=C(C(CC(CSC(C)=O)C(O)=O)=O)C=C1 | |
280.34 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.