missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4,6-dichloro-5-nitronicotinate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 154012-15-4 |
Assay Percent Range | 95% |
Molecular Formula | C8H6Cl2N2O4 |
MDL Number | MFCD09842620 |
Quantity | 1 g |
Synonym | ethyl 4,6-dichloro-5-nitronicotinate, 4,6-dichloro-5-nitronicotinic acid ethyl ester, ethyl4,6-dichloro-5-nitronicotinate, ethyl4,6-dichloro-5-nitro-3-pyridinecarboxlate, 4,6-dichloro-5-nitro-nicotinic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonyl-3-nitropyridine, ethyl 4,6-dichloro-5-nitro-3-pyridinecarboxylate |
InChI Key | VMNMZVKEDHEOEJ-UHFFFAOYSA-N |
SMILES | CCOC(=O)C1=CN=C(Cl)C(=C1Cl)[N+]([O-])=O |
IUPAC Name | ethyl 4,6-dichloro-5-nitropyridine-3-carboxylate |
Molecular Weight (g/mol) | 265.05 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethyl 4,6-dichloro-5-nitronicotinate, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction