missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 5-bromo-2-chlorobenzoate, 98+%, Thermo Scientific™ Product Code.: 11424113

Ethyl 5-bromo-2-chlorobenzoate, 98+%, Thermo Scientific™

Product Code. 11424113
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11424113

Brand: Thermo Scientific Alfa Aesar A10334.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76008-73-6
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.515
MDL Number MFCD00013635
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
PubChem CID 144749
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl

Specifications

Density 1.48
Boiling Point 156°C to 157°C (13 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 5 g
Beilstein 6501991
Formula Weight 263.52
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-bromo-2-chlorobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-bromo-2-chlorobenzoate, 98+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.