missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, for HPLC, Fisher Chemical™

Product Code. 11453383
2.5 L, Amber glass bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
MDL Number 9171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate

Specifications

Packaging Amber glass bottle
Quantity 2.5 L
Vapor Pressure 103 mbar at 20°C
Viscosity 0.45 cp at 20°C
Vapor Density 3.04
Physical Form Liquid
Color Colorless
Melting Point -83.5°C
Boiling Point 75°C
SMILES CCOC(C)=O
Molecular Weight (g/mol) 88.11
Formula Weight 88.11 g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl Acetate, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.