missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

27.40 EUR - 444.00 EUR

Chemical Identifiers

CAS 35553-92-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026897
InChI Key XXVVNHCWPHMLEZ-UHFFFAOYSA-N
Synonym ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl esterShow MoreShow Less
PubChem CID 2774971
IUPAC Name ethyl 2-(3-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=CC=C1)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10793223
SDS View Documents
Thermo Scientific Maybridge
BTB09251DA
1 g
27.40 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10437222
SDS View Documents
Thermo Scientific Maybridge
BTB09251EA
10 g
138.00 EUR
10g
Please to purchase this item. Need a web account? Register with us today!
10252532
SDS View Documents
Thermo Scientific Maybridge
BTB09251EE
50 g
444.00 EUR
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.