Learn More
Ethylphosphonic dichloride, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1066-50-8 |
---|---|
Molecular Formula | C2H5Cl2OP |
Molecular Weight (g/mol) | 146.93 |
MDL Number | MFCD00002073 |
InChI Key | OWGJXSYVHQEVHS-UHFFFAOYSA-N |
Synonym | ethylphosphonic dichloride, phosphonic dichloride, ethyl, ethylphosphoryl dichloride, ethyldichlorophosphite, aurora ka-1351, acmc-20akvx, ethylphosphonyl dichloride, ethyl phosphonic dichloride, dichloroethylphosphine oxide, 1-dichloro-phosphoryl-ethane |
PubChem CID | 70596 |
IUPAC Name | 1-dichlorophosphorylethane |
SMILES | CCP(Cl)(Cl)=O |
Description
Ethylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylphosphonic dichloride finds it uses in the preparation of: ethylphosphonic diisocyanate, 4-acetylphenyl isopropyl ethylphosphonate, 4-acetylphenyl phenyl ethylphosphonate and 4-acetylphenyl cyclohexyl ethylphosphonate.
Solubility
Soluble in chloroform and dichloromethane.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1066-50-8 | |
146.93 | |
OWGJXSYVHQEVHS-UHFFFAOYSA-N | |
70596 | |
CCP(Cl)(Cl)=O |
C2H5Cl2OP | |
MFCD00002073 | |
ethylphosphonic dichloride, phosphonic dichloride, ethyl, ethylphosphoryl dichloride, ethyldichlorophosphite, aurora ka-1351, acmc-20akvx, ethylphosphonyl dichloride, ethyl phosphonic dichloride, dichloroethylphosphine oxide, 1-dichloro-phosphoryl-ethane | |
1-dichlorophosphorylethane |
Specifications
1066-50-8 | |
71°C to 72°C (12 mmHg) | |
C2H5Cl2OP | |
MFCD00002073 | |
Moisture sensitive | |
Soluble in chloroform and dichloromethane. | |
CCP(Cl)(Cl)=O | |
146.93 | |
146.94 | |
Ethylphosphonic dichloride |
1.376 | |
>110°C (230°F) | |
1.4665 | |
UN2922 | |
ethylphosphonic dichloride, phosphonic dichloride, ethyl, ethylphosphoryl dichloride, ethyldichlorophosphite, aurora ka-1351, acmc-20akvx, ethylphosphonyl dichloride, ethyl phosphonic dichloride, dichloroethylphosphine oxide, 1-dichloro-phosphoryl-ethane | |
OWGJXSYVHQEVHS-UHFFFAOYSA-N | |
1-dichlorophosphorylethane | |
70596 | |
98% |
Safety and Handling
GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 213-918-3
missing translation for 'rtecsNumber' : TA1780000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.