Learn More
Invitrogen™ EVOS™ Light Cube, RFP
The revolutionary design of the EVOS™ light cubes combines a high intensity, long lifetime light source (LED), condensing optics, dichroic mirror and filters into a compact, adjustable, easy-to-install accessory for fluorescence detection on the EVOS™ imaging systems and the Countess II FL.
Brand: Invitrogen™ AMEP4652
This item is not returnable.
View return policy
Description
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
RFP Fluorescent Light Cube | |
Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System | |
Ex: 531/40 Em: 593/40 |
EVOS | |
LED, Condensing Optics, Hard-coated Filters | |
1 Ea. |
Application Note: Using time-lapse imaging with the EVOS FL Auto Imaging System
File size: 610.7 kB
Application Note - Collecting Z-stack Image Sequences
File size: 292.4 kB
Product Content Correction
Product Title
Invitrogen™ EVOS™ Light Cube, RFP > RFP
Spot an opportunity for improvement?Share a Content Correction