missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXD1 Monoclonal Antibody (OTI5H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502099
This item is not returnable.
View return policy
Description
EXD1 is an RNA-binding component of the PET complex, a multiprotein complex required for the processing of piRNAs during spermatogenesis. The piRNA metabolic process mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposable elements, preventing their mobilization, which is essential for the germline integrity. The PET complex is required during the secondary piRNAs metabolic process for the PIWIL2 slicing-triggered loading of PIWIL4 piRNAs. In the PET complex, EXD1 probably acts as an RNA adapter. EXD1 is an inactive exonuclease.Specifications
| EXD1 | |
| Monoclonal | |
| 0.81 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NHP7 | |
| EXD1 | |
| Full length human recombit protein of human EXD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI5H1 | |
| Unconjugated | |
| EXD1 | |
| EXDL1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 161829 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction