missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome Component 9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Bio-Techne NBP1-71702SS
This item is not returnable.
View return policy
Description
Exosome Component 9 Polyclonal specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Exosome Component 9 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa | |
Rabbit | |
Affinity Purified | |
RUO | |
5393 | |
Human, Mouse, Chicken | |
IgG |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
0.6 mg/mL | |
Western Blot 1:5000, Simple Western 1:1000, Chromatin Immunoprecipitation reported in scientific literature (PMID 27543448), Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:50-1:100, Chromatin Immunoprecipitation (ChIP) | |
Q06265, Q9JHI7 | |
EXOSC9 | |
A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265] | |
0.025 mL | |
Primary | |
Human, Mouse and Chicken. Immunogen has 83% identity to rat and 90% identity to bovine. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Exosome Component 9 Antibody - BSA Free, Novus Biologicals™ > 0.025mL; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction