Learn More
FA2H Monoclonal Antibody (OTI2E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506817
Encompass_Preferred
 View more versions of this product
Product Code. 17556352
  • 540.87 EUR / 100µL
Description
FA2H is a protein that catalyzes the synthesis of 2-hydroxysphingolipids, a subset of sphingolipids that contain 2-hydroxy fatty acids. Sphingolipids play roles in many cellular processes and their structural diversity arises from modification of the hydrophobic ceramide moiety, such as by 2-hydroxylation of the N-acyl chain, and the existence of many different head groups.
Specifications
Specifications
FA2H
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q7L5A8
FA2H
Full length human recombit protein of human FA2H produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2E3
Unconjugated
FA2H
FAAH, FAH1, FAXDC1, SCS7, SPG35
Mouse
Affinity Chromatography
RUO
79152
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
FA2H Monoclonal Antibody (OTI2E3), TrueMAB™, OriGene