Learn More
FA2H Monoclonal Antibody (OTI2E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506817S

Description
FA2H Monoclonal antibody specifically detects FA2H in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
FA2H | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P0DML2 | |
Mouse | |
Affinity chromatography | |
RUO | |
1442 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI2E3 | |
Unconjugated | |
CSH1 | |
FA2H; FAAH; FAH1; Fatty acid 2-hydroxylase; fatty acid alpha-hydroxylase; fatty acid hydroxylase domain containing 1; Faxdc1; FLJ25287; G630055L08Rik; RGD1310347; SCS7; spastic paraplegia 35 (autosomal recessive); SPG35; WD repeat domain 59; Wdr59 | |
Full length human recombinant protein of human CSH1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.