Learn More
BD Biosciences™ FACSCount Control Kit (CE/IVD)
For setting up the BD FACSCount™ instrument and for checking linearity.
Brand: BD Biosciences™ 340166
Description
- Linearity checking.
- Four bead concentrations (zero, low, medium, and high).
- Contained in two tube pairs with color-coded tops.
- Pair one:
- Zero (yellow top): 0 beads/μL
- Low (red top): 50 beads/μL
- Pair two:
- Medium (blue top): 250 beads/μL
- High (purple top): 1,000 beads/μL

Specifications
Low, Medium, and High bead: 2.0 x 107 to 3.0 x 107/each | |
In-vitro Diagnostics, FACSCount Instrument Linearity Check | |
BD FACSCount™ | |
25 Runs |
Suitable for 25 runs | |
FACSCount Control Kit | |
Four bead concentrations (Zero, Low, Medium, and High) | |
Low bead: approx. 50 beads/μL, Medium bead: approx. 250 beads/μL, High bead: approx. 1,000 beads/μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.