Learn More
Fadrozole hydrochloride, MedChemExpress
MedChemExpress Fadrozole hydrochloride (CGS 16949A) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.
Brand: Medchem Express HY-14247

Description
Fadrozole hydrochloride (CGS 16949A) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.Specifications
Fadrozole hydrochloride | |
White | |
C14H14ClN3 | |
CGS 16949A(Rac)-FAD286 hydrochloride | |
N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1.Cl | |
259.73 | |
Research | |
Cancer-programmed cell death |
102676-31-3 | |
Research | |
100 mg | |
DMSO : 100 mg/mL (385.02 mM; Need ultrasonic and warming) ∣H2O : 100 mg/mL (385.02 mM; Need ultrasonic) | |
259.73 | |
98.11% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only