Learn More
Falcon™ Pipet Controller Replacement Parts
Assortment of replacement parts for pipet controllers.
4.91 EUR - 83.44 EUR
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
15379840
![]() |
Falcon™
357475 |
Casing part 1 and 2 |
46.66 EUR
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15359840
![]() |
Falcon™
357474 |
Silicone pipet holder |
25.05 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15389840
![]() |
Falcon™
357476 |
Nosecone |
5.97 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15399840
![]() |
Falcon™
357477 |
Connector gasket |
7.27 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15329850
![]() |
Falcon™
357479 |
Operation buttons |
21.06 EUR
Pack of 2 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15319850
![]() |
Falcon™
357478 |
Connector |
5.30 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15339850
![]() |
Falcon™
357480 |
Divider casing |
24.51 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15349850
![]() |
Falcon™
357481 |
Spring |
4.91 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15369850
![]() |
Falcon™
357482 |
O-ring |
12.63 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15379850
![]() |
Falcon™
357483 |
Silcone tubes |
18.91 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15399850
![]() |
Falcon™
357485 |
Pump |
69.95 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15389850
![]() |
Falcon™
357484 |
Control board |
83.44 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15319860
![]() |
Falcon™
357486 |
Battery |
29.18 EUR
Pack of 3 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15339860
![]() |
Falcon™
357487 |
Battery Cover (U.S. version) |
21.06 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15349860
![]() |
Falcon™
357488 |
Battery Cover (EU version) |
21.06 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15379860
![]() |
Falcon™
357490 |
2-position charging stand |
49.56 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
15359860
![]() |
Falcon™
357489 |
Universal Power Supply |
40.81 EUR
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
Description
For use with Falcon pipet controller.Specifications
Pipet controller |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Falcon™ Pipet Controller Replacement Parts