Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Brand: Value Plastics 50120-49
This item is not returnable.
View return policy
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtimeSpecifications
Non-DEHP;Non-Phthalate;RoHS | |
Nylon | |
Female Luer x Hose Barb |
1/16 in. | |
25/Pk. |
Product Content Correction
Product Title
Value Plastics Female Luer to 200 Series Barb > 1/16 in.,Nylon
Always test the application to verify the performance of the interconnection.
Spot an opportunity for improvement?Share a Content Correction