Learn More
Thermo Scientific™ TranscriptAid T7 High Yield Transcription Kit
Get high yield in vitro transcription from DNA templates containing a T7 RNA Polymerase promoter, including linearized plasmids or PCR products.
Brand:  Thermo Scientific™ K0441
447.45 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10818990
  • 476.00 EUR / 50 reactions
Description
TranscriptAid T7 High Yield Transcription Kit contains reagents for 50 reactions of 20 μL. Each reaction yields up to 200 μg RNA from 1 μg of template in 2 hours (see Figures 1 and 2). The reaction can be scaled up to produce milligram amounts of full-length RNA. The kit can be used to produce both long and short transcripts for applications that require large yields of RNA (see Figure 3). All necessary reagents for transcription are included, as well as the RiboRuler High Range RNA Ladder for sizing and quantification.
Highlights include:
  • Exceptionally high yields—up to 200 μg in 2 hours
  • Versatile—suitable for both short and long RNA transcripts
  • Milligram amounts of RNA in a single, scaled-up reaction
  • Flexible—generates unlabeled, labeled or capped RNA
  • RiboRuler RNA Ladder supplied with kit for sizing and quantification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TranscriptAid
TranscriptAid T7 High Yield Transcription Kit
50 reactions
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2796633 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0441
Lot Number2796711 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0441
Lot Number2797154 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0441
Lot Number2796519 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0441
Lot Number2827871 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0441
Special Offers
Special Offers
  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977
    Expires2025-05-31
Product Content Correction
Product Title
Thermo Scientific™ TranscriptAid T7 High Yield Transcription Kit > for 50 rxn of 20uL