missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroportin/SLC40A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 EUR - 596.00 EUR
Specifications
Antigen | Ferroportin/SLC40A1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1.0 ug/ml, Flow Cytometry 1.0 ug/ml. Use reported by customer review, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787453
|
Novus Biologicals
NBP1-21502SS |
0.025 mL |
292.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18720074
|
Novus Biologicals
NBP1-21502 |
0.1 mL |
596.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferroportin/SLC40A1 Polyclonal specifically detects Ferroportin/SLC40A1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Ferroportin/SLC40A1 | |
Western Blot 1.0 ug/ml, Flow Cytometry 1.0 ug/ml. Use reported by customer review, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) | |
Polyclonal | |
Rabbit | |
Cancer, Lipid and Metabolism, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
SLC40A1 | |
IgG | |
Affinity Purified | |
62.5 kDa |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Pig, Bovine | |
Q9NP59 | |
30061 | |
This Ferroportin/SLC40A1 Antibody was developed against a synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-300). [Swiss-Prot: Q9NP59] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ferroportin/SLC40A1 Antibody - BSA Free, Novus Biologicals™