Learn More
Ferroportin/SLC40A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-21502
 View more versions of this product
Product Code. 18720074
  • 596.00 EUR / 0.10mL
Explore more special offers
Description
Description
Ferroportin/SLC40A1 Polyclonal specifically detects Ferroportin/SLC40A1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferroportin/SLC40A1
Rabbit
Human, Mouse, Pig, Bovine
Western Blot, Immunohistochemistry (Paraffin)
0.1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ferroportin/SLC40A1 Antibody - BSA Free, Novus Biologicals™ > 0.1mL; Unlabeled