missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF21 Monoclonal Antibody (OTI9A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502850

This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Specifications
FGF21 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NSA1 | |
FGF21 | |
Human recombit protein fragment corresponding to amino acids 29-209 of human FGF21 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI9A7 | |
Unconjugated | |
FGF21 | |
fibroblast growth factor 21 | |
Mouse | |
Affinity Chromatography | |
RUO | |
26291 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
FGF21 Monoclonal Antibody (OTI9A7), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction