Learn More
Eurofins DiscoverX FGFR1 (V561M) Protein, Active
Active, N-terminal GST-tagged, recombinant, human FGFR1 amino acids 456 - 765 containing the V561 mutation. The amino acid valine 561 was mutated to a methionine in FGFR1 which corresponded to previously reported mutations found in Abl (T315) and EGFR (T766) which had been shown to confer resistance to selective inhibitors, for use in Enzyme Assays.
Brand: Eurofins DiscoverX 14-734M

Description
Fibroblast growth factor (FGF) superfamily consists of 23 members, all of which contain a conserved 120 amino acid (aa) core region that contains six identical, interspersed amino acids. Based on the pI factor, two main groups of FGF are named: acidic FGF (aFGF) and basic FGF (bFGF). The superfamily members act extracellularly through four tyrosine kinase FGF receptors (FGFR1 through FGFR4), with multiple specificities noted for almost all FGFs. Biological binding of aFGF to its receptors will result in phosphorylation of the receptor and non-receptor proteins and generate its function. Overexpression and amplification of the fibroblast growth factor receptor-1 (FGFR-1), a prototypic receptor tyrosine kinase, have been found in tumor samples from human pancreas, breast, brain, prostate, thyroid gland, and salivary gland.
GenBank NM_000604
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥79% |
62.3 kDa | |
Human | |
N-terminal GST | |
Truncated | |
FGFR1 (V561M), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.