Learn More
Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes
Use these fixed volume single-channel pipettes for highly demanding applications where robustness and durability are essential.
Brand: Thermo Scientific™ 4652010
135.20 EUR valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Use Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, these pipettes are built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 | |
Autoclavable | |
1 | |
5 μL |
FinnpipetteTM F2 fixed volume | |
PVDF | |
Fixed Volume Pipette |
Accessories and Services
-
Thermo Scientific™ Finntip™ Non-Filtered 384 Format P...
X1000 Pipettor tip 0.2µL to 20µL Finntip... Product Code: 10152532122.00 EUR / Pack of 1000 -
Thermo Scientific™ Finntip™ Pipette Tips
X3840 PIPETTE TIP FINNTIP 50µL, RACK 0.2... Product Code: 10423322632.00 EUR / Pack of 3840 -
Thermo Scientific™ Finnpipette™ Stands
PIPETTE STAND 6 POSITIONS Product Code: 11455748142.00 EUR / Each -
Thermo Scientific™ Finntip™ Flex Filtered Pipette Tip...
X960 Filtered Finntip Flex 10ul, sterile... Product Code: 11833430275.00 EUR / Pack of 960 -
Thermo Scientific™ Finntip™ Flex™ Pipette Tips
X960 Finntip Flex 10ul, Non Sterile, rac... Product Code: 11863420149.00 EUR / Pack of 960 -
Thermo Scientific™ Finntip™ Flex™ Pipette Tips
X960 Tips Finntip Flex rackedpolypropyle... Product Code: 11873420171.00 EUR / Pack of 960 -
Thermo Scientific™ Finntip™ Flex™ Pipette Tips X2016 Tips Finntip Flex racked polypropy... Product Code: 11873430
172.15 EUR / 1 set -
Thermo Scientific™ Finntip™ Flex™ Pipette Tips
X1000 Tips Finntip Flex polypropylene Ba... Product Code: 11883420101.30 EUR / Pack of 1000 -
Thermo Scientific™ Finntip™ Flex™ Pipette Tips
X1920 Tips Finntip Flex racked polypropy... Product Code: 11883430221.00 EUR / Pack of 1920
Special Offers
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13