Learn More
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Adapters connect electrodes to various input jacks and are mercury free
Brand: Fisherbrand™ 01X559301
This item is not returnable.
View return policy
Description
Adapters connect electrodes to various input jacks and are mercury free
- Adapters connect electrodes to various input jacks
Specifications
US Standard/Pin Plug to BNC | |
Electrode | |
Connection Adapter | |
Adapts electrode with BNC plug to U.S. Standard/Pin input jack |