Learn More
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Adapters connect electrodes to various input jacks and are mercury free
133.00 EUR - 167.00 EUR
Specifications
For Use With (Equipment) Electrode
Type Connection Adapter
View More Specs
Products 3
Product Code Brand Connector Types For Use With (Application) Price Quantity & Availability  
Product Code Brand Connector Types For Use With (Application) Price Quantity & Availability  
11540124
View Documents Promotion Details
Fisherbrand™
01X559301
US Standard/Pin Plug to BNC Adapts electrode with BNC plug to U.S. Standard/Pin input jack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11550124
View Documents Promotion Details
Fisherbrand™
13620493
US Standard/Pin Plug to Radiometer Type 7 Adapts electrode with U.S. Standard/Pin plugs to Radiometer Type 7 input jack
133.00 EUR
Each
Estimated Shipment: 21-11-2024
to see stock.
Add to basket
11560124
View Documents Promotion Details
Fisherbrand™
01X559501
US Standard/Pin Plug to BNC dapts electrode with U.S. Standard/Pin plugs to BNC input jack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Adapters connect electrodes to various input jacks and are mercury free
  • Adapters connect electrodes to various input jacks
Specifications
Specifications
Electrode
Connection Adapter