Learn More
Fisherbrand™ Electrode Connection Adapters - Mercury - Free
Adapters connect electrodes to various input jacks and are mercury free
Brand: Fisherbrand™ 13620493
This item is not returnable.
View return policy
Description
Adapters connect electrodes to various input jacks and are mercury free
- Adapters connect electrodes to various input jacks
Specifications
US Standard/Pin Plug to Radiometer Type 7 | |
Electrode | |
Connection Adapter | |
Adapts electrode with U.S. Standard/Pin plugs to Radiometer Type 7 input jack |