Learn More
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 2mm Thick
For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.
Brand:  Fisherbrand™ MS7-2-2
 View more versions of this product
Product Code. 11867553
  • 21.40 EUR / Each
Estimated Shipment: 27-06-2025
Description
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximises sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Blue
Comb
1 Each
180 ÎĽL
Sub-Gel Mini Horizontal Gel Electrophoresis Unit
2 Prep, 2 Marker
2 mm
Videos
  • Fisherbrand and Fisher Bioreagents - Focus on electrophoresis
    Fisherbrand provides a comprehensive range of horizontal gel systems for all your nucleic acid gel electrophoresis needs.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand and Fisher BioReagents Focus on electrophoresis
    Product Title
    Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 2mm Thick >