Learn More
Fisherbrand™ Lens Paper Product Code.: 15392267
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
Fisherbrand™ Lens Paper
Product Code. 15392267
6 x 8 in. sheets, 100 Sheets/Book, 12 Books/Pk.
Click to view available options
Dimensions (L x W):
4 x 6 in.
6 x 8 in. sheets
Quantity:
100 Sheets/Book, 12 Books/Pk.
50 Sheets/Book, 12 Books/Pk.
Product Code. 15392267
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
Add to basket
For cleaning glass lenses
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Dispenser Box
  • Same paper as above, but packaged in a convenient dispenser box for fast, single-sheet removal individual sheets measure 5L × 7 in.W (13 × 18cm)
Specifications
Product Type Lens Cleaning Paper
Dimensions (L x W) 6 x 8 in. sheets
For Use With (Application) For cleaning Glass lenses
Quantity 100 Sheets/Book, 12 Books/Pk.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2126880 Certificate TypeCertificate of Analysis Date07/03/2020 Catalog Number11995
Lot Number2124951 Certificate TypeCertificate of Analysis Date07/03/2020 Catalog Number11995
Lot Number2126865 Certificate TypeCertificate of Analysis Date04/03/2020 Catalog Number11995
Lot Number2124887 Certificate TypeCertificate of Analysis Date04/03/2020 Catalog Number11995
Lot Number2116896 Certificate TypeCertificate of Analysis Date02/03/2020 Catalog Number11995
Product Content Correction
Product Title
Fisherbrand™ Lens Paper >