Learn More
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps Product Code.: 15331129
Estimated Shipment: 16-04-2026
Only null left
Add to basket
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
GREENER_CHOICE
Product Code. 15331129 Shop All Fisherbrand Products
Natural, 1.5 mL, 18,000 x g, Skirted, Pack of 500
Click to view available options
Color:
Natural
Capacity (Metric):
0.5 mL
1.5 mL
Max. RCF:
18,000 x g
30,000 x g
Bottom Shape:
Skirted
Unit Size:
Pack of 500
Product Code. 15331129
Only null left
Estimated Shipment: 16-04-2026
Only null left
Add to basket
Add to basket
Choose skirted- or conical-bottom style.
  • Threaded to accept colored screw caps (sold separately)
  • Siliconized/low-retention conical-bottom tubes also available
Note: For caps, see Fisherbrand™ Colored Screw Caps. 
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1.5 mL
Color Natural
Material Plastic
Sterility Non-sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Skirted
Max. RCF 18,000 x g
Product Content Correction
Product Title
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps >