Learn More
Fisherbrand™ Polypropylene Tubing Equal Angle Y-Shaped Connectors
Highly resistant to many chemicals.
Brand:  Fisherbrand™ 1531531A
 View more versions of this product
Product Code. 16660554
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Excellent chemical resistance
  • Autoclavable polypropylene
  • Serrated connecting surfaces to prevent tubing slipping
  • Withstands flows to 20 p.s.i.
  • The elasticity of the tubing will influence choice of size
  • Dimensions listed are nominal
Specifications
Specifications
Equal Angle Tubing Connector
0.125 to 0.219 in.
1.4 in.
4 to 6mm
0.2 in.
30.48 mm
White
Autoclavable
Polypropylene
35.5 mm
5.08 mm
Y
1.2 in.
20 psi