Learn More
Fisherbrand™ Acid Resistant Plastic Tubs for Ultrasonic Cleaners
Provide additional tank protection when using harsh chemical cleaning solutions in the ultrasonic cleaner.
54.35 EUR - 246.00 EUR
Product Code | Brand | For Use With (Equipment) | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Capacity (Metric) | Price | Quantity & Availability | |||||
10248761
|
Fisherbrand™
2000008000 |
Ultrasonic Cleaner, S 300/S 300 H |
246.00 EUR
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
10177381
|
Fisherbrand™
2070900000 |
Ultrasonic Cleaners: S 100, S 100/H; S 120, S 120/H | 5.8 L |
159.00 EUR
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
10320892
|
Fisherbrand™
2070890000 |
Ultrasonic Cleaners: S 30, S 30/H; S 40, S 40/H; S 60, S 60/H | 1.3 L |
54.35 EUR
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
10137431
|
Fisherbrand™
2070910000 |
Ultrasonic Cleaners: S 180, S 180/H | 11.0 L |
211.00 EUR
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
Description
- Designed to provide additional protection against harsh chemical cleaning solutions
- Comes with a custom-fit cover to prevent leaks and spills
Specifications
Acid Resistant Tub | |
Plastic cover | |
Plastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Acid Resistant Plastic Tubs for Ultrasonic Cleaners