Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1003285
 View more versions of this product
Product Code. 10487912
  • 37.95 EUR / Each
Estimated Shipment: 09-06-2025
Description
  • Easy-to-open and close cover
  • Muffles noise level
  • Designed to be used as drip basin when flipped over
  • Chemical-resistant plastic
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage
Specifications
Specifications
Lid
Ultrasonic Cleaner, 9.5 and 12.75 L
Medical Laboratories and in Industrial Production Processes
Plastic
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 4:02
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:02
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
SDS
Documents
Documents
Product Certifications
Product Title
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners > For use with FB11207 Ultrasonic Unit