Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Brand: Thermo Scientific™ R112641
183.02 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
- For sterility testing according to USP (United States Pharmacopeia)
Specifications
Industrial Reference: AOAC, BAM, COMPF, USP. | |
United States | |
Microbiology Media |
SB | |
Fluid Thioglycollate Medium | |
10/Pk. |
Product Content Correction
Product Title
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium > SB - 100mL; 10/Pk.
Spot an opportunity for improvement?Share a Content Correction