Learn More
Memmert™ Flush-Fit Sets without Air Slots for SF450
Stainless steel frame covering gap between oven and wall opening.
Brand: Memmert™ SF450B29741
This item is not returnable.
View return policy
Description
- Stainless steel
- Without air slots
- Techincal clarification required
Specifications
SF450 Sterilizer | |
Stainless Steel |
Product Content Correction
Product Title
Memmert™ Flush-Fit Sets without Air Slots for SF450 > For Use With: SF450
Spot an opportunity for improvement?Share a Content Correction