Learn More
Honeywell™ FORCE (i)XTREM S3 Shoes
Honeywell™ FORCE (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
Brand: Honeywell™ 6246203-45/7
This item is not returnable.
View return policy
Description
- Water resistant grain full leather
- Ergonomic collar and padded tongue for an optimized comfort
- Gusset to avoid water and dust penetration
- Breathable 3D lining
- Derby style for a better fitting in all sizes
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Specifications
EN ISO 20345:2011 S3 HRO HI CI SRC | |
Leather | |
Polyurethane and nitrile |
Black | |
45 | |
Metal-free |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Honeywell™ FORCE (i)XTREM S3 Shoes > Size: 45
Spot an opportunity for improvement?Share a Content Correction