Learn More
BRAND™ Forceps
PTFE forceps
Brand:  BRAND™ 113130
48.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10746861
Description
Description
  • Rectangular ends
  • Outstanding chemical and thermal resistance
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Rectangular
    PTFE
    150 mm
    1
    Documents
    One moment while we fetch your results.
    Product Title
    BRAND™ Forceps > Length (Metric): 150 mm