missing translation for 'onlineSavingsMsg'
Learn More
Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Honeywell Fluka™ Product Code.: 15602830

Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Honeywell Fluka™
BIOPHARMA

Product Code. 15602830 Shop All Honeywell Fluka Products
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
70 kg
1200 kg
This item is not returnable. View return policy

Product Code. 15602830

Brand: Honeywell-Fluka 270012.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specifications of DAC, FCC, 98.0-100%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow MoreShow Less
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Linear Formula HCOOH
UN Number 1779
Beilstein 1209246
Formula Weight 46.03g/mol
Percent Purity 98 to 100%
Grade puriss.
Density 1.22g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.57
Flash Point 48°C (118.4°F)
Ignition Point 1004°F
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8mmHg (20°C)
Chemical Name or Material Formic Acid, Meets analytical specifications of DAC, FCC
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM070A Certificate TypeCertificate of Analysis Date14/06/2023 Catalog Number27001-2.5L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.