Learn More
Cytiva Funnel Housing
Replacement housing for funnels with unique magnetic seal.
Brand:  Cytiva 4248
Product Code. 17194281
  • 353.00 EUR / Each
Estimated Shipment: 28-07-2025
Description
  • Magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyphenylsulfone
Non-sterile
150 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1423208 Certificate TypeCertificate of Processing Date21/03/2025 Catalog Number4248
Lot Number1417170 Certificate TypeCertificate of Processing Date12/02/2025 Catalog Number4248
Product Title
Cytiva Funnel Housing > 150 mL