Learn More
Furfuryl mercaptan, 97%, Thermo Scientific Chemicals
47.64 EUR - 122.61 EUR
Chemical Identifiers
CAS | 98-02-2 |
---|---|
Molecular Formula | C5H6OS |
Molecular Weight (g/mol) | 114.16 |
MDL Number | MFCD00003254 |
InChI Key | ZFFTZDQKIXPDAF-UHFFFAOYSA-N |
Synonym | furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol |
PubChem CID | 7363 |
IUPAC Name | furan-2-ylmethanethiol |
SMILES | C1=COC(=C1)CS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636692
|
Thermo Scientific Acros
119810250 |
25 mL | Glass bottle |
47.64 EUR
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377080
|
Thermo Scientific
119811000 |
100 mL | Glass bottle |
122.61 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-02-2 | |
114.16 | |
ZFFTZDQKIXPDAF-UHFFFAOYSA-N | |
7363 | |
C1=COC(=C1)CS |
C5H6OS | |
MFCD00003254 | |
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol | |
furan-2-ylmethanethiol |
Specifications
98-02-2 | |
1.1300g/mL | |
45°C | |
96% min. (GC) | |
1.5290 to 1.5330 | |
17, II, 116 | |
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol | |
ZFFTZDQKIXPDAF-UHFFFAOYSA-N | |
furan-2-ylmethanethiol | |
7363 | |
97% | |
Furfuryl mercaptan, 97% |
Colorless to Yellow | |
155.0°C | |
Authentic | |
C5H6OS | |
MFCD00003254 | |
1.13 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in chloroform | |
C1=COC(=C1)CS | |
114.16 | |
114.16 | |
Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-628-2
missing translation for 'rtecsNumber' : LU2100000
missing translation for 'tsca' : TSCA
- Furfuryl mercaptan
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.