Learn More
Fisherbrand™ Gamma Radiation Safe Mini Storage Box
Provides radiation protection shielding for use in laboratories.
Brand: Fisherbrand™ CSR-B0.4G
Description
- Manufactured with hinged lids
- Transparent shielding
- Serves as solution for short-term storage of radioactive waste or radioisotopes
- Constructed of acrylic provides easy fabrication and manufacturability
Specifications
8.5 x 8.5 x 5.5cm (3.35 x 3.35 x 2.16 in.) | |
Radioisotopes or radioactive waste | |
Mini Storage Box |
Gamma radiation shielded | |
Hinged lid | |
Acrylic |