Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Brand: Thermo Scientific™ 290OA241
30.45 EUR valid until 2025-04-30
Use promo code "24211" to get your promotional price.
Description
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
Viton Liner Sealing Ring | |
10 Pack | |
Viton Liner Sealing Ring | |
TRACE 1300 Series GC and Agilent SSL Injectors |
Special Offers
-
Multiple Manufacturers
Chromatography Monthly Promotion
Discover exclusive monthly deals on chromatography supplies to boost your lab's efficiency.Promo Code: 24211Expires: 2025-04-30
New Offer Save Up to 67%