Learn More
Lonza™ GelBond™ Film for Agarose Gels
Transparent, flexible polyester film supports gels through electrophoresis, immunodiffusion, fixing, staining, destaining, and drying procedures. GelBond™ Film for Agarose Gels are reliable, durable and convenient.
Brand:  Lonza™ 53750
 View more versions of this product
Product Code. 11557755
  • 1675.00 EUR / 1 roll
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Description
  • Permanently attaches to gels during casting
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining and drying
  • Available as precut sheets or rolls
Note: polyester films will not transmit light of less than 310nm and will fluoresce at higher wavelengths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose Gel Film
Polyester
0.007 in.
649.6 in.
5 in.
Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures
1 Roll
0.02 cm
16.5 m
12.7 cm