Learn More
Lonza™ GelBond™ Film for Agarose Gels
Transparent, flexible polyester film supports gels through electrophoresis, immunodiffusion, fixing, staining, destaining, and drying procedures. GelBond™ Film for Agarose Gels are reliable, durable and convenient.
Brand:  Lonza™ 53759
 View more versions of this product
Product Code. 11567755
  • 2300.00 EUR / 100 sheets
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Description
  • Permanently attaches to gels during casting
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining and drying
  • Available as precut sheets or rolls
Note:polyester films will not transmit light of less than 310nm and will fluoresce at higher wavelengths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose Gel Film
Polyester
0.007 in.
9.6 in.
4.9 in.
Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures
100 Sheets
0.02 cm
24.5 cm
12.5 cm