Learn More
Lonza™ GelBond™ Film for Agarose Gels
Transparent, flexible polyester film supports gels through electrophoresis, immunodiffusion, fixing, staining, destaining, and drying procedures. GelBond™ Film for Agarose Gels are reliable, durable and convenient.
Brand: Lonza™ 53759
This item is not returnable.
View return policy
Description
- Permanently attaches to gels during casting
- Makes gels easier to handle
- Helps gels retain their original dimensions during staining and drying
- Available as precut sheets or rolls
Note: polyester films will not transmit light of less than 310nm and will fluoresce at higher wavelengths
Specifications
Agarose Gel Film | |
Polyester | |
0.007 in. | |
9.6 in. | |
4.9 in. |
Agarose Gel electrophoresis, immunodiffusion, fixing, staining, destaining and drying procedures | |
100 Sheets | |
0.02 cm | |
24.5 cm | |
12.5 cm |