Learn More
Gemcitabine elaidate, MedChemExpress
MedChemExpress Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.
Brand: Medchem Express HY-13538
This item is not returnable.
View return policy
Description
Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.Specifications
Gemcitabine elaidate | |
White | |
C27H43F2N3O5 | |
CP-4126 CO-101 Gemcitabine 5'-elaidate | |
O[C@@H](C(F)(F)[C@H](N1C(N=C(C=C1)N)=O)O2)[C@H]2COC(CCCCCCC/C=C/CCCCCCCC)=O | |
527.64 | |
Research | |
Cancer-Kinase/protease |
210829-30-4 | |
Research | |
50 mg | |
DMSO : ≥ 26 mg/mL (49.28 mM) | |
527.64 | |
97.42% | |
Solid |