Learn More
Gemcitabine elaidate, MedChemExpress
MedChemExpress Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.
Brand:  Medchem Express HY-13538
Encompass_Preferred
 View more versions of this product
Product Code. 16421907
  • 544.05 EUR / 50mg
Description
Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.
Specifications
Specifications
Gemcitabine elaidate
White
C27H43F2N3O5
CP-4126 CO-101 Gemcitabine 5'-elaidate
O[C@@H](C(F)(F)[C@H](N1C(N=C(C=C1)N)=O)O2)[C@H]2COC(CCCCCCC/C=C/CCCCCCCC)=O
527.64
Research
Cancer-Kinase/protease
210829-30-4
Research
50 mg
DMSO : ≥ 26 mg/mL (49.28 mM)
527.64
97.42%
Solid