Learn More
Gemcitabine elaidate, MedChemExpress
MedChemExpress Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.
120.15 EUR - 544.05 EUR
Specifications
Chemical Name or Material | Gemcitabine elaidate |
---|---|
CAS | 210829-30-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C27H43F2N3O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16438427
|
Medchem Express
HY-13538 |
5 mg |
120.15 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421907
|
Medchem Express
HY-13538 |
50 mg |
544.05 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16415748
|
Medchem Express
HY-13538 |
10 mg |
172.80 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gemcitabine elaidate (CP-4126) is lipophilic pro-drug of Gemcitabine. Gemcitabine elaidate is converted to Gemcitabine by esterases in order to be phosphorylated. Gemcitabine elaidate exhibits anti-tumor activity.Specifications
Gemcitabine elaidate | |
White | |
C27H43F2N3O5 | |
DMSO : ≥ 26 mg/mL (49.28 mM) | |
527.64 | |
97.42% | |
Solid |
210829-30-4 | |
Research | |
CP-4126 CO-101 Gemcitabine 5'-elaidate | |
O[C@@H](C(F)(F)[C@H](N1C(N=C(C=C1)N)=O)O2)[C@H]2COC(CCCCCCC/C=C/CCCCCCCC)=O | |
527.64 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.