missing translation for 'onlineSavingsMsg'
Learn More
Gestodene, Thermo Scientific™ Product Code.: 17352491

Gestodene, Thermo Scientific™

Product Code. 17352491
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 17352491

Brand: Thermo Scientific Acros 468650010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60282-87-3
Molecular Formula C21H26O2
Molecular Weight (g/mol) 310.44
MDL Number 00867858
InChI Key SIGSPDASOTUPFS-KQMXEUTGSA-N
Synonym 13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one
IUPAC Name (1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C

Specifications

Packaging Glass Bottle
Quantity 1 g
Certificate of Analysis Available
Physical Form Powder
Merck Index 4425
Color White to Beige
Melting Point 197.9°C
Formula Weight 310.43
Chemical Name or Material Gestodene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gestodene, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.