Learn More
Ginkgolide A, MedChemExpress
MedChemExpress Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.
Brand: Medchem Express HY-B0355
This item is not returnable.
View return policy
Description
Ginkgolide A (BN-52020) is an extract from in Ginkgo biloba and a g-aminobutyric acid (GABA) antagonist.Specifications
Ginkgolide A | |
White | |
C20H24O9 | |
BN-52020 | |
O=C1[C@@H](C)[C@]2(O)[C@@]3(C4=O)C(C5([C@@H](O)C(O6)=O)C6O3)([C@H](O4)C[C@H]5C(C)(C)C)C[C@]2([H])O1 | |
408.4 | |
Research | |
COVID-19-immunoregulation |
15291-75-5 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (244.86 mM; Need ultrasonic) | |
408.4 | |
98.0% | |
Solid |